missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYBR™ Fast SYBR™ Green Master Mix
Designed for speed, SYBR™ Green Master Mix provides a fast, reliable, and cost-effective solution for RT-PCR application without compromising sensitivity, specificity, dynamic range, or PCR efficiency.
£106.00 - £3535.00
Specifications
Detection Method | SYBR |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Fast DNA Polymerase, AmpliTaq Fast UP |
Reaction Speed | Fast |
Includes: SYBR Green I Dye, AmpliTaq Gold Fast DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference I and optimized buffer components
Requires: Template and primers
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10381275
|
SYBR™
4385610 |
1 x 1 mL |
£106.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10459604
|
SYBR™
4385612 |
1 x 5 mL |
£434.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10556555
|
SYBR™
4385616 |
2 x 5 mL |
£768.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10566555
|
SYBR™
4385617 |
5 x 5 mL |
£1828.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10631376
|
SYBR™
4385618 |
10 x 5 mL |
£3535.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10447494
|
SYBR™
4385614 |
1 x 50 mL |
£3390.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- A fast, reliable, and cost-effective solution for real-time PCR applications without compromising sensitivity, specificity, dynamic range, or PCR efficiency
- Real-time PCR results in as fast as 35 minutes
- Capable of detecting very low copies of target
- Minimizes primer-dimer and non-specific amplification
- Consistent amplification across a wide dynamic range
- Contains all of the components, excluding the template and primers, in a convenient 2X master mix
- Store at -5°C to -30°C
Includes the following components in an optimized buffer:
- AmpliTaq™ Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start
- Minimizes non-specific product formation and enables reactions to be set up at room temperature
- SYBR™ Green I dye to enable detection of double-stranded DNA
- Deoxynucleotides (dNTPs) to help maintain optimal PCR results
- Uracil-DNA Glycosylase (UDG) designed to reduce carryover contamination
- Passive internal reference based on proprietary ROX™ dye to enable increased precision
Order Info
Shipping Condition: Dry Ice
Specifications
SYBR | |
qPCR | |
Fast | |
SYBR Green | |
SYBR™ | |
UP (Ultra Pure) | |
2X |
High | |
AmpliTaq Fast DNA Polymerase, AmpliTaq Fast UP | |
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.