missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SYBR™ Green Universal Master Mix

Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.

Brand:  Applied Biosystems™ 4364346

 View more versions of this product

Product Code. 10177204


Includes: 2X mixture of SYBR Green I Dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components.
This item is not returnable. View return policy

Description

Description

Applied Biosystems™ SYBR™ Green Universal Master Mix combines SYBR™ Green I dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.

  • SYBR™ Green I dye detects double-stranded DNA, so specific probes are not required
  • AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
  • dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
  • Proprietary buffer enhancements ensure performance and reliability
  • Premixed components stored at 2° to 8°C significantly reduce assay setup time

Maximum Flexibility and Convenience:

  • Applied Biosystems™ SYBR™ Green Universal Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan™ probes are required
  • SYBR™ Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR
  • The hot-start enzyme AmpliTaq Gold™ DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity
  • Passive Internal Reference 1 is provided to normalize non-PCR—related fluorescence fluctuations
  • This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation
  • SYBR™ Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.
  • Store at -20°C

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ SYBR™ Green Universal Master Mix > 5 x 5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.