missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SYBR™ Green Universal Master Mix

Product Code. 10187094 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 5 mL
5 x 5 mL
10 x 5 mL
1 x 50 mL
Unit Size:
1 set
5 ampoules
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10187094

Brand: Applied Biosystems™ 4309155

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

2X mixture of SYBR Green I Dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components.

Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.

Applied Biosystems™ SYBR™ Green Universal Master Mix combines SYBR™ Green I dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.

  • SYBR™ Green I dye detects double-stranded DNA, so specific probes are not required
  • AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
  • dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
  • Proprietary buffer enhancements ensure performance and reliability
  • Premixed components stored at 2° to 8°C significantly reduce assay setup time

Maximum Flexibility and Convenience:

  • Applied Biosystems™ SYBR™ Green Universal Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan™ probes are required
  • SYBR™ Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR
  • The hot-start enzyme AmpliTaq Gold™ DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity
  • Passive Internal Reference 1 is provided to normalize non-PCR—related fluorescence fluctuations
  • This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation
  • SYBR™ Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.
  • Store at -20°C

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage The 1 x 5 mL vials contain a 2X mixture of SYBR Green 1 Dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components. Sufficient reagents provided for 200 reactions based on a 50 μl reaction volume.

Store at -20°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
Label or Dye SYBR Green
Passive Reference Dye ROX (Pre-mixed)
Product Line SYBR
Product Type Real Time PCR SYBR Master Mix
Quantity 1 x 5 mL
Shipping Condition Wet Ice
Sufficient For 200 Reactions of 50 μL
For Use With (Application) Gene Expression
No. of Reactions 200 Reactions
Sample Type DNA (Genomic), cDNA
Volume 1 x 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.