Learn More
Applied Biosystems™ SYBR™ Green Universal Master Mix
Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.
Brand: Applied Biosystems™ 4312704
2722.29 GBP valid until 2025-12-31
Use promo code "24271" to get your promotional price.
Includes: 2X mixture of SYBR Green I Dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components.
Description
Applied Biosystems™ SYBR™ Green Universal Master Mix combines SYBR™ Green I dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.
- SYBR™ Green I dye detects double-stranded DNA, so specific probes are not required
- AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
- dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
- Proprietary buffer enhancements ensure performance and reliability
- Premixed components stored at 2° to 8°C significantly reduce assay setup time
Maximum Flexibility and Convenience:
- Applied Biosystems™ SYBR™ Green Universal Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan™ probes are required
- SYBR™ Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR
- The hot-start enzyme AmpliTaq Gold™ DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity
- Passive Internal Reference 1 is provided to normalize non-PCR—related fluorescence fluctuations
- This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation
- SYBR™ Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.
- Store at -20°C
Order Info
Shipping Condition: Wet Ice

Specifications
The 10 x 5 mL vials contain a 2X mixture of SYBR Green 1 Dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components. Sufficient reagents provided for 2000 reactions based on a 50 μl reaction volume. Store at -20°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Tube | |
qPCR | |
Standard | |
SYBR Green | |
SYBR™ | |
10 x 5 mL | |
Gene Expression | |
2000 Reactions | |
10 x 5 mL |
SYBR | |
High | |
AmpliTaq Gold DNA Polymerase | |
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
2000 Reactions of 50 μL | |
2X | |
DNA (Genomic), cDNA |
Safety and Handling
- Green PCR Master Mix
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
Supplemental information
- MIXTURE LIST-Contains : Tris (hydroxymethyl)aminomethane, Sodium azide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.