missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TAMRA™ Dye Calibration Plate, Fast 96-well

Used to maintain your real-time PCR instrument with Fast 96-well block.

Brand:  Applied Biosystems™ 4432632

Product Code. 15860403

  • £167.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dye calibration should be run after lamp change. Please see your instrument user guide for full instructions on use.

Dye calibration should be run after lamp change. See instrument user guide for full instructions.

Order Info

Dry ice shipping; store in freezer (-5° to -30°C)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at –20°C ± 5°C.

ViiA™ 7 Dx System, QuantStudio™ Dx
Real Time PCR (qPCR)
1 x 96-well plate
TAMRA™, TAMRA™
Dye Calibration Plate
Dry Ice
Spectral Calibration
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For in vitro Diagnostic Use.