missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ TaqMan™ Control Genomic DNA (human)

Product Code. 10249144 Shop All Applied Biosystems Products
Click to view available options
Quantity:
2 Tubes
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10249144

Brand: TaqMan™ 4312660

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Conveniently packaged in a PCR-ready concentration for use as a control template in PCR reactions

The Human gDNA (male) is provided in two tubes (100μL each) at a concentration of 10ng/μL. TaqMan™ Control Genomic Human DNA is also included in several Applied Biosystems™ real-time PCR kits.

Order Info

Shipping Condition: Dry Ice

Recommended Storage

Store at -15° to -25°C.

TRUSTED_SUSTAINABILITY

Specifications

Format Frozen
Content And Storage Human gDNA (Male) packaged in two tubes (100μL each). Store at -15 to 25°C.
Species Human
Shipping Condition Dry Ice
Concentration 10 ng/μL
Label or Dye Unlabeled
Quantity 2 Tubes
Product Line TaqMan

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.