missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Fast Advanced Master Mix for qPCR

TaqMan Fast Advanced Master Mix delivers accurate quantification and dependability in less time on an array of qPCR instrument platforms. It provides best in class performance including the widest quantifiable range, in both single and duplex reactions, even with challenging targets.

Brand:  Applied Biosystems™ 4444557

331.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.



Code : 50

Additional Details : Weight : 0.02000kg

Product Code. 11380912

  • £382.00 / Each
In Stock
to see stock.

Description

Description

  • Delivers accurate quantification and dependability in less time on an array of qPCR instrument platforms
  • Provides best in class performance including the widest quantifiable range, in both single and duplex reactions, even with challenging targets
  • TaqMan Fast Advanced Master Mix contains AmpliTaq Fast DNA Polymerase, uracil-N-glycosylase (UNG), dNTPs with dUTP, ROX dye (passive reference), and optimized buffer components
  • Designed to provide performance equal to or better than standard master mixes
  • Engineered to retain a high level of performance in preassembled reactions for up to 72 hours
  • It is supplied at a 2X concentration
  • Store at 4°C

Features of the TaqMan Fast Advanced Master Mix include:

  • Best-in-class performance – superior sensitivity, accuracy, dynamic range, and specificity compared to standard mixes in standard mode
  • Engineered for enhanced benchtop stability – stable at room temperature for up to 72 hours in preassembled reactions
  • Optimized for multiplexing – validated for duplexing with exogenous and endogenous internal positive control assays
  • Reduced run times – optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamlessly transitions into experiments – validated with TaqMan assays for gene expression and microRNAs, and TaqMan array micro-fluidic cards

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

Contains 1 tube (5 ml) 2X TaqMan™ Fast Advanced Master Mix sufficient for 500 reactions

TaqMan™ Fast Advanced Master Mix contains AmpliTaq™ Fast DNA Polymerase, Uracil-N glycosylase (UNG), dNTPs (with dUTP), ROX™ dye (passive reference), and optimized buffer.

Store at 4°C.
Liquid
qPCR
Fast
TaqMan™
1 x 5 mL
Gene Expression, miRNA analysis, Virus Detection
2 X
DNA (Genomic), cDNA
Primer-probe
High
AmpliTaq Fast DNA Polymerase
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 6 Pro, QuantStudio™ 7 Flex, QuantStudio™ 7 Pro, StepOne™, StepOnePlus™, ViiA™ 7 System
Fast Advanced Master Mix
Wet Ice
2X
500 Reactions
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.