Learn More
Applied Biosystems™ TaqMan™ Fast Advanced Master Mix for qPCR
TaqMan Fast Advanced Master Mix delivers accurate quantification and dependability in less time on an array of qPCR instrument platforms. It provides best in class performance including the widest quantifiable range, in both single and duplex reactions, even with challenging targets.
Brand: Applied Biosystems™ 4444558
2260.11 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
- Delivers accurate quantification and dependability in less time on an array of qPCR instrument platforms
- Provides best in class performance including the widest quantifiable range, in both single and duplex reactions, even with challenging targets
- TaqMan Fast Advanced Master Mix contains AmpliTaq Fast DNA Polymerase, uracil-N-glycosylase (UNG), dNTPs with dUTP, ROX dye (passive reference), and optimized buffer components
- Designed to provide performance equal to or better than standard master mixes
- Engineered to retain a high level of performance in preassembled reactions for up to 72 hours
- It is supplied at a 2X concentration
- Store at 4°C
Features of the TaqMan Fast Advanced Master Mix include:
- Best-in-class performance – superior sensitivity, accuracy, dynamic range, and specificity compared to standard mixes in standard mode
- Engineered for enhanced benchtop stability – stable at room temperature for up to 72 hours in preassembled reactions
- Optimized for multiplexing – validated for duplexing with exogenous and endogenous internal positive control assays
- Reduced run times – optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
- Seamlessly transitions into experiments – validated with TaqMan assays for gene expression and microRNAs, and TaqMan array micro-fluidic cards
Order Info
Shipping Conditions: Dry Ice
Specifications
Contains 1 tube (50 mL) 2X TaqMan Fast Advanced Master Mix sufficient for 5000 reactions TaqMan Fast Advanced Master Mix contains AmpliTaq Fast DNA Polymerase, Uracil-N glycosylase (UNG), dNTPs (with dUTP), ROX dye (passive reference), and optimized buffer. Store at 4°C. |
|
Liquid | |
qPCR | |
Fast | |
TaqMan™ | |
1 x 50 mL | |
Gene Expression, miRNA analysis, Virus Detection | |
2 X | |
DNA (Genomic), cDNA |
Primer-probe | |
High | |
AmpliTaq Fast DNA Polymerase | |
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 6 Pro, QuantStudio™ 7 Flex, QuantStudio™ 7 Pro, StepOne™, StepOnePlus™, ViiA™ 7 System | |
Fast Advanced Master Mix | |
Wet Ice | |
2X | |
5000 reactions | |
50 mL |
For Research Use Only. Not for use in diagnostic procedures.