Learn More
TaqMan™ TaqMan™ Gene Expression Master Mix
Precise and reliable real-time qPCR quantification over 9 logs of linear dynamic range with Applied Biosystems™ TaqMan™ Gene Expression Master Mix.
Brand: TaqMan™ 4370048
43.38 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
TaqMan™ Gene Expression Master Mix has been validated with TaqMan™ Gene Expression Assays and Applied Biosystems™ real-time systems, providing precise quantification for a variety of gene expression-based real-time qPCR applications, including rare transcript detection, gene expression analysis, pathogen detection and viral load quantification,co-amplification of two targets, as well as specific detection of gene family members
TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure) for hot start activation and improved detection of bacterial targets, a blend of dNTPs with dTTP/dUTP and Uracil-DNA Glycosylase (UDG) to minimize carry-over PCR contamination, and a passive internal reference based on proprietary ROX™ dye for superb precision on Applied Biosystems™ real-time PCR instruments.
- Provides precise and reliable real-time qPCR quantification over nine logs of linear dynamic range
- TaqMan Gene Expression Master Mix is an optimized 2X mix that contains all of the components, excluding the template and primers, for sensitive detection down to one copy of target
- Extended benchtop stability provides consistent results for automated liquid-handling systems
- This makes it the choice for high-throughput sample processing overnight or over the weekend
- Validated with TaqMan gene expression assays and Applied Biosystems real-time systems, providing precise quantification for a variety of gene expression–based real-time qPCR applications
- Applications include rare transcript detection, gene expression analysis, pathogen detection and viral load quantification, co-amplification of two targets, as well as specific detection of gene family members
- Store at 2°C to 8°C
TaqMan Gene Expression Master Mix contains:
- AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), for hot start activation and improved detection of bacterial targets
- A blend of dNTPs with dTTP/dUTP and uracil-DNA glycosylase (UDG) to minimize carry-over PCR contamination
- A passive internal reference based on proprietary ROX dye for superb precision on Applied Biosystems real-time PCR instruments
Order Info
Shipping Condition: Wet ice
Specifications
1 tube (1 mL) 2X TaqMan™ Gene Expression Master Mix sufficient for 40 reactions. Store at 2°C to 8°C. TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase (Ultra Pure), Uracil-DNA glycosylase, dNTPs (with dUTP), ROX™ Passive Reference, and optimized buffer components. |
|
Liquid | |
AmpliTaq Gold DNA Polymerase | |
7000 System, 7300 System, 7500 System, 7900HT System, Applied Biosystems StepOnePlus™ Fast Real-Time PCR SystemStepOne™, Standard Mode, StepOnePlus™, Standard Mode | |
TaqMan™ | |
1 x 1 mL | |
40 Reactions | |
1 x 1 mL |
Primer-probe | |
qPCR | |
Standard | |
ROX (Pre-mixed) | |
UP (Ultra Pure) | |
Gene Expression | |
DNA (Genomic), cDNA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.