missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TaqMan™, GTXpress™ TaqMan™ GTXpress™ Master Mix
Combines AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye, and ROX™ dye in a convenient single vial format.
£104.00 - £6910.00
Specifications
GC-Rich PCR Performance | High |
---|---|
PCR Method | qPCR |
For Use With (Application) | Genotyping |
Concentration | 2X |
Includes: Contains 2 x 50mL TaqMan™ GTXpress™ Master Mix (2X master mix containing AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye and ROX™ Dye).
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10635975
|
Applied Biosystems™
4403311 |
1 mL |
£104.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10239234
|
Applied Biosystems™
4401892 |
10 mL |
£854.00
10mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10625975
|
Applied Biosystems™
4401890 |
50 mL |
£3435.00
20000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10605805
|
Applied Biosystems™
4401857 |
2 x 50 mL |
£6910.00
40000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TaqMan™ GTXpress™ Master Mix delivers accurate genotyping results with robust performance in less than 50 min. The 100mL pack (2 x 50mL) includes reagents sufficient to perform 40,000 PCR reactions (5μL/reaction).
- Compatible with both Fast and Standard mode thermal cycling conditions
- Pre-mixed components stored at 2° to 8°C significantly reduce assay setup time
- Validated with all types of Applied Biosystems™ TaqMan™ Genotyping Assays
- Extended PCR cycling can be used for poorly amplified or sample-limited reactions The TaqMan™ GTXpress™ Master Mix is also sold as part of TaqMan™ Sample-to-SNP™ Kit, which provides real-time PCR-ready DNA in 5 min. from a wide variety of samples, ranging from blood to buccal swabs to plant tissues. For more information on the TaqMan™ Sample-to-SNP™ Kit, refer to the Applied Biosystems™ TaqMan™ Sample-to-SNP™ Kit Protocol.
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
High | |
qPCR | |
Genotyping | |
2X |
For Research Use Only. Not for use in diagnostic procedures.