missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Multiplex Master Mix

Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorter run times (less than 40 minutes).

Brand:  Applied Biosystems™ 4486295

 View more versions of this product

Product Code. 15420624

  • £4660.00 / 50mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Optimized for multiplexing - validated for quadplex gene expression and genotyping assays
  • Best-in-class performance - superior sensitivity, accuracy, dynamic range, and specificity
  • Reduced run times on fast and standard instrumentation - optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamless integration into your experiments - validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
  • Carry-over contamination control - contains heat-labile UDG
  • Broad instrument compatibility
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 1 bottle (50 mL) of 2X TaqMan™ Multiplex Master Mix sufficient for 5000 reactions.


TaqMan™ Multiplex Master Mix contains AmpliTaq™ Fast DNA Polymerase, uracil-N glycosylase (UNG), dNTPs (with dUTP), MUSTANG PURPLE™ dye (passive reference), and optimized buffer.

Store in refrigerator (2–8°C).

High
AmpliTaq Fast DNA Polymerase
7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
Multiplex Master Mix
Gene Expression, Virus Detection
2 X
DNA
Primer-probe
Multiplex qPCR
Fast
TaqMan™
50 mL
2X
5000 Reactions
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.