missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Multiplex Master Mix

Product Code. 15441823 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 mL
5 mL
2 x 5 mL
50 mL
5 x 5 mL
10 x 5 mL
Unit Size:
1mL
50mL
5mL
Pack of 10
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 15441823

Brand: Applied Biosystems™ 4461884

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorter run times (less than 40 minutes).

  • Optimized for multiplexing - validated for quadplex gene expression and genotyping assays
  • Best-in-class performance - superior sensitivity, accuracy, dynamic range, and specificity
  • Reduced run times on fast and standard instrumentation - optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamless integration into your experiments - validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
  • Carry-over contamination control - contains heat-labile UDG
  • Broad instrument compatibility
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains 2 bottles (5 mL each) of 2X TaqMan™ Multiplex Master Mix sufficient for 1000 reactions.


TaqMan™ Multiplex Master Mix contains AmpliTaq™ Fast DNA Polymerase, uracil-N glycosylase (UNG), dNTPs (with dUTP), MUSTANG PURPLE™ dye (passive reference), and optimized buffer.

Store in refrigerator (2–8°C).

Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method Multiplex qPCR
Polymerase AmpliTaq Fast DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
Product Line TaqMan
Product Type Multiplex Master Mix
Quantity 2 x 5 mL
Sufficient For 1000 Reactions
For Use With (Application) Gene Expression, Virus Detection
Fidelity (vs. Taq) 2 X
No. of Reactions 1000 Reactions
Sample Type DNA
Volume 10 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.