missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Multiplex Master Mix
Alle ansehen Applied Biosystems Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
5 mL
2 x 5 mL
50 mL
5 x 5 mL
10 x 5 mL
missing translation for 'unitSize'
1mL
50mL
5mL
Pack of 10
Pack of 2
Pack of 5
Beschreibung
- Optimized for multiplexing - validated for quadplex gene expression and genotyping assays
- Best-in-class performance - superior sensitivity, accuracy, dynamic range, and specificity
- Reduced run times on fast and standard instrumentation - optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
- Seamless integration into your experiments - validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
- Carry-over contamination control - contains heat-labile UDG
- Broad instrument compatibility
Spezifikation
Spezifikation
| Concentration | 2X |
| Content And Storage | Contains 1 bottle (5 mL) of 2X TaqMan™ Multiplex Master Mix sufficient for 500 reactions. TaqMan™ Multiplex Master Mix contains AmpliTaq™ Fast DNA Polymerase, uracil-N glycosylase (UNG), dNTPs (with dUTP), MUSTANG PURPLE™ dye (passive reference), and optimized buffer. Store in refrigerator (2–8°C). |
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | High |
| PCR Method | Multiplex qPCR |
| Polymerase | AmpliTaq Fast DNA Polymerase |
| Reaction Speed | Fast |
| For Use With (Equipment) | 7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System |
| Product Line | TaqMan |
| Product Type | Multiplex Master Mix |
| Mehr anzeigen |
For Research Use Only. Not for use in diagnostic procedures.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur