Learn More
Applied Biosystems™ TaqMan™ Reagents Starter Kit
TaqMan™ PCR Starter Kit is designed to detect and quantify Eukaryotic 18S rRNA sequence in human Raji cDNA using the TaqMan™ 5' nuclease assay with the TaqMan™ PCR Master Mix.
Brand: Applied Biosystems™
219.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.50000kg
Includes: TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG, Eukaryotic 18S rRNA TaqMan™ Assay (with two unlabeled PCR primers and a labeled TaqMan™ probe), plus a sample of human Raji cDNA
Description
Gene-specific, ready to use TaqMan™ Gene Expression Assays and Universal Master Mix provide you with a fast, simple way for measuring changes in gene expression.
- Simple procedure gives you quick results
- Hot-Start DNA Polymerase enzyme system minimizes nonspecific product amplification for reliable results
- Used With: 7500 Real-Time PCR System with Dell™ Notebook, 7500 Real-Time PCR System with Dell™ Tower, 7900HT Fast Real-Time PCR System with 384-well Block Module, 7900HT Fast Real-Time PCR System with 384-well Block Module and Automation Accessory, 7900HT Fast Real-Time PCR System with Standard 96-well Block Module, 7900HT System Automation Accessory Upgrade, 7900HT System Standard 384-well Block Upgrade Kit, 7900HT System Standard 96-well Block Upgrade Kit, ABI PRISM™ 7000 Sequence Detection System (Notebook)
Ready-to-Use Gene Expression Solution:
- The TaqMan™ Reagents Starter Kit contains a sample of TaqMan™ Universal PCR Master Mix, and TaqMan™ Gene-Expression Assay (Eukaryotic 18S rRNA), plus a sample of human Raji cDNA - everything you need for 18S gene-expression analysis
- Simply combine these reagents according to the instructions to generate reproducible and quantitative gene-expression data
A Complete, Integrated Gene-Expression System:
- Eliminate time-consuming master mix formulation and assay optimization with the TaqMan™ Gene-Expression Assays and the TaqMan™ Universal PCR Master Mix, by using universal cycling conditions and pre-optimized reagents
Order Info
Shipping Conditions: Dry Ice
Specifications
This kit contains TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG, Eukaryotic 18S rRNA TaqMan™ Assay (with two unlabeled PCR primers and a labeled TaqMan™ probe), plus a sample of human Raji cDNA. Store the PCR mix at 2° to 8°C. Store the rest of the kit contents at -15° to -25°C. |
|
qPCR | |
Standard | |
ROX (Pre-mixed) | |
PCR Reagents | |
18S rRNA | |
Gene Expression | |
DNA (Genomic), cDNA |
Primer-probe | |
AmpliTaq Gold DNA Polymerase | |
7000 System, 7500 System, AB 7900 HT 384-Well System | |
TaqMan™ | |
Dry Ice | |
Target-Specific | |
Built-In Hot Start |
For Research Use Only. Not for use in diagnostic procedures.