Learn More
Applied Biosystems™ TaqMan™ RNase P Instrument Verification Plate, Fast 48-well
Used to verify the performance of the Applied Biosystems™ StepOne™ Real-Time PCR System.
Brand: Applied Biosystems™ 4371439
237.60 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Includes: Contains 48-well Fast Reaction Plate preloaded and sealed with TaqMan™ RNase P probes and primers.
Description
The TaqMan™ RNase P Fast 48-well Instrument Verification Plate is a 48-well PCR plate preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template. The TaqMan™ RNase P Fast 48-well Instrument Verification Plate satisfies the same requirements as the Applied Biosystems™ StepOne™ Real-Time PCR Systems installation. To pass installation, the instrument must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems™ recommends that the performance of your instrument be verified every six months.
The TaqMan RNase P Fast 48-Well Instrument Verification Plate satisfies the same requirements as the Applied Biosystems StepOne Real-Time PCR Systems installation. To pass installation, the instrument must demonstrate the ability to distinguish between 5000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, the performance of the instrument should be verified every six months.
Order Info
Dry ice shipping; store at -15° to 25°C; product is light sensitive
Specifications
Contains 48-Well Fast Reaction Plate preloaded and sealed with TaqMan™ RNase P probes and primers. Store at -15 to 25°C. This product is light sensitive. | |
StepOne™, Fast Mode | |
TaqMan™ | |
Real Time PCR (qPCR) | |
1 x 48-well plate |
FAM | |
Fluorescence | |
Instrument Verification Plate | |
Dry Ice | |
RNase P Calibration, Instrument Verification Plate |
For Research Use Only. Not for use in diagnostic procedures.