Learn More
Applied Biosystems™ Total RNA Control (Human)
Provided at a convenient reverse-transcription-ready concentration for all RT-PCR reactions
Marke: Applied Biosystems™ 4307281
Code : 50
Weitere Details : Weight : 0.10500kg
Beschreibung
TaqMan™ Control Total RNA is from Raji cells (Burkitt's lymphoma), and is conveniently packaged in a PCR-ready concentration for use as a control template in RT-PCR reactions. The Human total RNA is provided at a concentration of 50ng/μL.
Bestellinformation
Shipping Condition: Dry Ice
Spezifikation
Frozen | |
Human | |
50 ng/μL |
100 μl Total Human RNA (at 50 ng/μl). Store at -15 to -25°C. | |
Dry Ice | |
Unlabeled |
For Research Use Only. Not for use in diagnostic procedures.