Learn More
Applied Biosystems™ VeritiLink™ Remote Management Software
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 software
Unit Size:
Each
Description
VeritiLink Remote Management Software (VRMS) browser-enabled interface means you can “Bookmark Your Veritis” for fast, convenient access
- Create and edit methods on your computer for increased security and peace of mind, and download/upload to/from your Veriti systems as needed
- Perform and monitor runs for up to 50+ networked Veriti Thermal Cyclers
- Manage all Veriti™ systems in secure client-server environment with user password protection
- Receive real-time email system updates on desktop, laptop or PDA
- Easy setup: install software on server for use on any networked computers
- Veriti Systems Anywhere, Everywhere: VeritiLink Remote Monitoring Software VRMS allows you to access Veriti Systems anywhere, everywhere
- Software package allows to manage and monitor thermal cycling runs wherever you can access an internet browser
- With this program, you can start or stop runs, create and edit methods and send them to Veriti Thermal Cycler systems as needed
- Designed to manage from one to 50+ Veriti Thermal Cyclers, user interface shows which systems are available for use and status of systems in use to facilitate instrument sharing for busy labs
- With system update email notifications to your desktop, laptop, or PDA and user password protection to keep your methods secure
- Software provides with peace of mind that systems are running smoothly and securely
- Easy to Install: Software is easy to install, requiring only server-side installation to provide access for all networked computers
PCR, PCR & Real-Time PCR
Specifications
Specifications
| For Use With (Equipment) | Veriti Thermal Cycler |
| Product Line | VeritiLink |
| Software Type | Remote Management Software |
| Product Type | Thermal Cycler Accessories |
| For Use With (Application) | PCR |
| Quantity | 1 software |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido