missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Trays and Dividers for Shell Vials
Accommodate narrow- or wide-diameter vials.
£201.00
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
10149701
|
Fisherbrand™
AS261 |
Tray for Wide Vials | For wide vials (28.5 x 95 mm) |
£201.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Cardboard trays have cell dividers
- 10 x 10 in. (25 x 25cm) row configuration
Specifications
Not Autoclavable | |
Cardboard | |
Dividers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title