Learn More
IBA Solutions For Life Science™ ApU-dinucleotide IBA
Dinucleotides are used by RNA polymerases as initiators for RNA synthesis. Therefore, they are very useful for transcription analysis.
Brand: IBA Solutions For Life Science™ 5-0710-147
Code : LE
Additional Details : Weight : 0.10000kg
Specifications
10 mg |