missing translation for 'onlineSavingsMsg'
Learn More

AQ-RA 741, Tocris Bioscience™

Product Code. 18035202
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective, high affinity M2 antagonist

High affinity, selective muscarinic M2 receptor antagonist (pKi values are 8.3, 7.7 and 6.82 for M2, M1 and M3 receptors, respectively). Displays cardioselectivity in vivo, over intestinal, tracheal and bladder muscarinic receptors; inhibits vagally and agonist-induced bradycardia.

Chemical Identifiers

CAS 123548-16-3
Molecular Formula C27H37N5O2
Molecular Weight (g/mol) 463.626
InChI Key BCUGCHZRMKTPMU-UHFFFAOYSA-N
Synonym aq-ra 741, aq-ra-741, 11-4-4-diethylamino butyl-1-piperidinyl acetyl-5,11-dihydro-6h-pyrido 2,3-b 1,4 benzodiazepin-6-one, 6h-pyrido 2,3-b 1,4 benzodiazepin-6-one,11-2-4-4-diethylamino butyl-1-piperidinyl acetyl-5,11-dihydro, acmc-20ej93, d0vz3d, aq-ra-741bs, chembl43383, 11-2-4-4-diethylamino butyl piperidin-1-yl acetyl-5h-benzo e pyrido 3,2-b 1,4 diazepin-6 11h-one
PubChem CID 129989
IUPAC Name 11-[2-[4-[4-(diethylamino)butyl]piperidin-1-yl]acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one
SMILES CCN(CC)CCCCC1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4

Specifications

Quantity 50 mg
Formula Weight 463.62
Percent Purity >99%
Chemical Name or Material AQ-RA 741
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'