missing translation for 'onlineSavingsMsg'
Learn More
AQ-RA 741, Tocris Bioscience™ Product Code.: 30001423

AQ-RA 741, Tocris Bioscience™

Product Code. 30001423
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001423

Brand: Tocris Bioscience 2292/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective, high affinity M2 antagonist

High affinity, selective muscarinic M2 receptor antagonist (pKi values are 8.3, 7.7 and 6.82 for M2, M1 and M3 receptors, respectively). Displays cardioselectivity in vivo, over intestinal, tracheal and bladder muscarinic receptors; inhibits vagally and agonist-induced bradycardia.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123548-16-3
Molecular Formula C27H37N5O2
Molecular Weight (g/mol) 463.626
InChI Key BCUGCHZRMKTPMU-UHFFFAOYSA-N
Synonym aq-ra 741, aq-ra-741, 11-4-4-diethylamino butyl-1-piperidinyl acetyl-5,11-dihydro-6h-pyrido 2,3-b 1,4 benzodiazepin-6-one, 6h-pyrido 2,3-b 1,4 benzodiazepin-6-one,11-2-4-4-diethylamino butyl-1-piperidinyl acetyl-5,11-dihydro, acmc-20ej93, d0vz3d, aq-ra-741bs, chembl43383, 11-2-4-4-diethylamino butyl piperidin-1-yl acetyl-5h-benzo e pyrido 3,2-b 1,4 diazepin-6 11h-one
PubChem CID 129989
IUPAC Name 11-[2-[4-[4-(diethylamino)butyl]piperidin-1-yl]acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one
SMILES CCN(CC)CCCCC1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4

Specifications

Quantity 10 mg
Formula Weight 463.62
Percent Purity >99%
Chemical Name or Material AQ-RA 741
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
AQ-RA 741, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.