Learn More
Macherey-Nagel™ Aquadur™ Test Strips
Determine water hardness quickly, easily and reliably. The Aquadur™ test strips provide a convenient dip and read procedure and a clear colour changes from green to red ensures reliable results within seconds.
Brand: Macherey-Nagel™ 91220
Code : 26
Additional Details : CAS Number : 7732-18-5 Weight : 0.03600kg
Description
- Quick and easy determination of water hardness.
- The convenient dip and read procedure and the clear colour changes from green to red ensure reliable results within seconds.
- Water hardness test strips come in a wide variety of different gradations and packaging options for your individual needs.
Specifications
CE, 93/42 / EEC | |
Determination of water hardness | |
100 Pack | |
Aquadur Test Strips |
Strip | |
<3 > 4 > 7 > 14 <21°d | |
20 sec. |