missing translation for 'onlineSavingsMsg'
Learn More

Aqualine™ Complete 2, Fisher Chemical™

Single-component reagent; Pyridine free

Brand:  Fisher Chemical K/1950/15

195.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 10744761

  • £262.00 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 2.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Brown
Amber glass bottle
81734
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Aqualine Complete 2

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Iodine, 1-Imidazole,Diethylene glycol monoethyl ether

SDS
Documents

Documents

Certificates
Special Offers

Special Offers