missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Electrolyte AD-G, for Karl Fischer Titration By Coulometry, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Chemical Name or Material | Aqualine™ Electrolyte ADG |
| Name Note | Karl Fisher Reagent |
| CAS | 7446-09-5, 108-75-8, 124-68-5 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Assay Percent Range | 50 to 75% |
| MDL Number | 81734 |
| Packaging | Amber glass bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction