missing translation for 'onlineSavingsMsg'
Learn More
Karl Fischer Aqualine™ Sodium Tartrate-2H2O, for Karl Fischer Titration, Fisher Chemical Product Code.: 10261552

Karl Fischer Aqualine™ Sodium Tartrate-2H2O, for Karl Fischer Titration, Fisher Chemical

Product Code. 10261552
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 10261552

Brand: Fisher Chemical K/2770/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solid standard

  • Primary solid water standard for Karl Fischer volumetric analysis
  • Contains 15.66% ± 0.05% water
TRUSTED_SUSTAINABILITY

Specifications

Color White
Quantity 100 g
CAS 6106-24-7
pH 7 to 9
Identification Pass Test
Packaging Plastic powder jar
Physical Form Powder/Solid
Molecular Formula C4H8Na2O8
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
Molecular Weight (g/mol) 230.08
PubChem CID 131855972
Formula Weight 230.08g/mol
Chemical Name or Material Aqualine™ Sodium Tartrate Dihydrate Water Standard, Karl Fischer Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Fischer Aqualine™ Sodium Tartrate-2H2O, for Karl Fischer Titration, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.