missing translation for 'onlineSavingsMsg'
Learn More
Karl Fischer Aqualine™ Water Standard 1.0mg, for Karl Fischer Titration, Fisher Chemical Product Code.: 10323102

Karl Fischer Aqualine™ Water Standard 1.0mg, for Karl Fischer Titration, Fisher Chemical

Product Code. 10323102
10 x 4 mL
Click to view available options
Quantity:
10 x 4 mL
This item is not returnable. View return policy

Product Code. 10323102

Brand: Fisher Chemical K/2710/99

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1mg/mL H2O standard

  • Standard contains 1mg H2O/mL (1000ppm)
  • Used for titer determination of coulometric or volumetric Karl Fischer reagents
  • Supplied as 10 x 4mL and packaged in glass ampules to protect from moisture
  • The ampule should only be opened immediately prior to use to prevent absorption of moisture from atmosphere
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Quantity 10 x 4 mL
CAS 7732-18-5, 105-13-5, 100-66-3
pH 7
Equivalence 0.97 to 1.03mg of water/mL
Packaging Ampoule
Physical Form Liquid
MDL Number 240423
Chemical Name or Material Aqualine™ Standard 1.1, Karl Fischer Reagent
compliance-icons compliance-icons
Product Identifier
  • Aqualine Water Standard 1 mg/g
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Fischer Aqualine™ Water Standard 1.0mg, for Karl Fischer Titration, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.