missing translation for 'onlineSavingsMsg'
Learn More
Karl Fischer Aqualine™ Water Standard 10mg, for Karl Fischer Titration, Fisher Chemical Product Code.: 10546242

Karl Fischer Aqualine™ Water Standard 10mg, for Karl Fischer Titration, Fisher Chemical

Product Code. 10546242
10 x 8 mL
Click to view available options
Quantity:
10 x 8 mL
This item is not returnable. View return policy

Product Code. 10546242

Brand: Fisher Chemical K/2720/99

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

10mg/mL H2O standard

  • Standard contains 10mg H2O/mL
  • Used for titer determination of volumetric Karl Fischer reagents
  • Supplied as 10 x 8mL and packaged in glass ampules to protect from moisture
  • The ampule should only be opened immediately prior to use to prevent absorption of moisture from atmosphere
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 x 8 mL
CAS 71-36-3
Equivalence 9.9 to 10.1mg of water/mL
Packaging Ampoule
Physical Form Liquid
Molecular Formula C4H10O
MDL Number 240423
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILES CCCCO
IUPAC Name butan-1-ol
Molecular Weight (g/mol) 74.123
PubChem CID 263
ChEBI CHEBI:28885
Chemical Name or Material Aqualine™ Standard 10.1, Karl Fischer Reagent
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Aqualine Water Standard 10 mg/g
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Fischer Aqualine™ Water Standard 10mg, for Karl Fischer Titration, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.