Learn More
Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical
Brand: Fisher Chemical K/2200/17
188.00 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 24
Additional Details : CAS Number : 67-56-1 Weight : 2.50000kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Specifications
Colorless | |
Pyridine free | |
Karl Fischer Reagent | |
64°C | |
Liquid | |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol |
2.5 L | |
Aqualine™ Titrant 5 | |
5.0mg of water/mL min. | |
Amber glass bottle | |
> 1.0 | |
MFCD00004595 | |
CO | |
32.04 |