Learn More
MP Biomedicals™ AquaPhenol™
Water saturated phenol for DNA/RNA extractions
Brand: MP Biomedicals™ 11AQUAPH01
View more versions of this product
Description
Phenol is used as a reagent in chemical analysis. It is used in the manufacture of colorless or light-colored artificial resins, many medical and industrial organic compounds and dyes. It is commonly used in tissue preservation and DNA/RNA extraction with chloroform.Specifications
Store at +4°C. Store under nitrogen. Protect from light. | |
4.5 - 5 (Lit.) | |
250 mL |
Water saturated Phenol | |
Molecular Biology | |
Solution |
Safety and Handling
- AquaPhenol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Phenol, ultra pure
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.