missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ AquaPhenol™
Water saturated phenol for DNA/RNA extractions
Brand: MP Biomedicals™ 11AQUAPH02
View more versions of this product
This item is not returnable.
View return policy
Description
Phenol is used as a reagent in chemical analysis. It is used in the manufacture of colorless or light-colored artificial resins, many medical and industrial organic compounds and dyes. It is commonly used in tissue preservation and DNA/RNA extraction with chloroform.Specifications
Store at +4°C. Store under nitrogen. Protect from light. | |
4.5 - 5 (Lit.) | |
4 x 250 mL |
Water saturated Phenol | |
Molecular Biology | |
Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction