Learn More
Stuart™ Aquatron™ deionizer Housing and Accessories
Stuart™ Aquatron™ deionizer Housing and Accessories provide good, quality water at an affordable price
Brand:  Stuart™ 80244-95
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 16140921
  • £27.72 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Output up to 60 litres/hour
  • Low cost disposable cartridges
  • Colour change indicates resin condition at a glance
  • Free standing or wall mountable
  • Ideal as pre-treatment for Aquatron™ water stills
  • A simple, portable deionizer giving good quality water at an affordable price
  • The disposable ion exchange cartridges slowly change colour from green to blue as they are exhausted giving at a glance indication of the resin condition
  • No need for conductivity meters, batteries or mains electrical power
  • Supplied complete with flexible hose for connection to a tap and a stopcock to control the flow rate
  • Can be linked in series with the Aquatron™ filter either by a flexible hose or the rigid coupling (ALC) available as an accessory
  • Ideal as a pre-treatment for Aquatron™ water stills to prevent scale build up and improve output quality
  • Housing and ion exchange cartridges must be ordered separately
Specifications
Specifications
Aquatron Filter
Wall Bracket
Double