missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standards, 84μS/cm, Mettler Toledo™
Click to view available options
Quantity:
250 mL
Description
- Non-hazardous due to aqueous composition
- Reduced shipping, storage, handling or disposal problems which normally occur with solvent-based conductivity standards
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 250 mL |
Color | Colorless |
Odor | Odorless |
Vapor Pressure | 23hPa |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'