missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ AR-GLAS™ Serological Pipets
Manufactured from AR-GLAS™ Soda-lime glass. BRAND™ AR-GLAS™ Serological Pipets include a large tip diameter to allow for total delivery. Short graduation marks and inscriptions in ETERNA amber stain.
Brand: Brand™ 27115
12.08 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from AR-GLAS™ Soda-lime glass.
- Calibrated to deliver (TD, Ex).
- Large tip diameter allows for total delivery.
- Short graduation marks and inscriptions in ETERNA amber stain.
Specifications
To Deliver | |
AR-GLAS, Soda Lime Glass | |
0.1 mL | |
±0.4 mL | |
Serological Pipette |
2 mm | |
Yes | |
-5 mL | |
25 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction