missing translation for 'onlineSavingsMsg'
Learn More

Brand™ AR-GLAS™ Serological Pipets

Manufactured from AR-GLAS™ Soda-lime glass. BRAND™ AR-GLAS™ Serological Pipets include a large tip diameter to allow for total delivery. Short graduation marks and inscriptions in ETERNA amber stain.

Brand:  Brand™ 27107

5.51 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10394511

  • £96.80 / Pack of 12
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from AR-GLAS™ Soda-lime glass.
  • Calibrated to deliver (TD, Ex).
  • Large tip diameter allows for total delivery.
  • Short graduation marks and inscriptions in ETERNA amber stain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

To Deliver
AR-GLAS, Soda Lime Glass
0.01 mL
±0.02 mL
Serological Pipette
2 mm
Yes
-0.2 mL
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.