Learn More
Hirschmann™ AR-Glass Graduated Pipet
Provides volumetric accuracy for your pipetting needs. Hirschmann™ Clear Glass Graduated Pipettes with Amber Stain Graduation with a cotton plug end as standard, short form for tissue culture.
Brand:  Hirschmann™ 110015327
 View more versions of this product
Product Code. 11781516
  • £86.50 / Pack of 12
Explore more special offers
Description
Description
  • Main point ring graduation
  • Cotton plug end
  • Class AS, Conformity certified, with dated batch
Specifications
Specifications
A
Soda Lime Glass
Amber
Durable amber with silver diffusion color
0.02 mL
360 ±5 mm
Short form
Yes
0.05 mL
12 Pack
5 mL
Graduated Pipet