missing translation for 'onlineSavingsMsg'
Learn More

AR-M 1000390 hydrochloride, Tocris Bioscience™

Product Code. 18053822
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Low-internalizing δ agonist

Non-peptidic, low-internalizing δ-selective opioid receptor agonist; derivative of SNC 80 (Cat. No. 0764). Does not trigger acute desensitization of the analgesic response; reduces CFA-induced hyperalgesia. Brain penetrant following systemic administration.

Chemical Identifiers

CAS 209808-47-9
Molecular Formula C23H29ClN2O
Molecular Weight (g/mol) 384.948
InChI Key OTXTZCLQEGSAMW-UHFFFAOYSA-N
Synonym ar-m 1000390 hydrochloride, arm390 hydrochloride, arm-390hcl, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl-benzamide hydrochloride, n,n-diethyl-4-phenyl piperidin-4-ylidene methyl benzamide hydrochloride, arm1000390 hydrochloride, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl benzamide hydrochloride
PubChem CID 76848958
IUPAC Name N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide;hydrochloride
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)C(=C2CCNCC2)C3=CC=CC=C3.Cl

Specifications

Quantity 10 mg
Formula Weight 384.94
Percent Purity >97%
Chemical Name or Material AR-M 1000390 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'