missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AR-M 1000390 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Non-peptidic, low-internalizing δ-selective opioid receptor agonist; derivative of SNC 80 (Cat. No. 0764). Does not trigger acute desensitization of the analgesic response; reduces CFA-induced hyperalgesia. Brain penetrant following systemic administration.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 384.94 |
Percent Purity | >97% |
Chemical Name or Material | AR-M 1000390 hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'