Learn More
Hecht Karl™ Automatic Burettes
Designed with improved special pinch cock with titration screw for precise regulation. VITLab™ Automatic Burettes are standard type with 1.5 times the tolerance of Class AS.
Brand: Hecht Karl™ 40820025
This item is not returnable.
View return policy
Description
- Schilling burettes
- Plastic burette clamp, chemical-resistant, allows discharge drop by drop by slightly pushing
- Polyethylene storage bottles, plastic base
- Burette fills by slightly pressing the storage bottle displaying automatic zero point adjustment
- Plastic tubing as riser
- Upper part of the burette is closed, Pellet titration burette design avoids soiling
- Burette with Schellbach stripe and fused graduation “ASSISTENT-PERMANENT”, calibrated, faceted outlet tip
Specifications
25 mL | |
0.1 mL | |
±0.1 mL |
Soda Lime Glass | |
Plastic Filling Tube, Polyethylene Bottle with Plastic Base Burette | |
Automatic Buret |