missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Aracut™ Grip Gloves
9.79 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Use promo code "21640" to get your promotional price.
Click to view available options
Size:
6
7
11
Description
- Knitted Kevlar™ glove.
- Hand conformity and good dexterity.
- Blue PVC dots on palm for grip and abrasion resistance.
- Knitted construction for hand conformity and dexterity.
- Excellent cutting resistance and protection against thermal hazards in a dry atmosphere.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-94 EEC Basic standards, EN 388-94 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/05/08/0134, EC Attestation Number 0075/797/162/05/08/0134 |
Coated Area | Palm, Fingertips |
Description | Gauge 10 |
Coating Material | PVC Dots |
Color | Yellow |
Cut Resistance Rating | 3 |
Material | Aramid Fiber, Dupont™ Kevlar™ Aramid Fibers |
Product Type | Gloves |
Size | 7 |
For Use With (Application) | Automotive, Building, Construction, Glass, Metal and Paper Industries |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'