missing translation for 'onlineSavingsMsg'
Learn More
Honeywell™ ARATHERMA FIT Gloves Product Code.: 12605343
In Stock
to see stock.
Add to basket

Honeywell™ ARATHERMA FIT Gloves

Product Code. 12605343
Size: 7
Click to view available options
Size: 7

Product Code. 12605343

Brand: Honeywell™ 223208607

In Stock
to see stock.
Add to basket
Add to basket

Honeywell™ ARATHERMA FIT Gloves for the handling of hot, sharp parts in a dry atmosphere.

  • 100% Kevlar™ knitted glove
  • Lined with one cotton knitted glove
  • Long knitted elastic wrist (10 cm)
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • Kevlar™ glove lined with cotton is pleasant to the touch
  • The lining enhances the glove's mechanical performance while absorbing perspiration
  • The glove is ambidextrous and thus more cost-effective
  • The glove's relative thickness (gauge 7) enables it to absorb shocks, making it suitable for the handling of heavy objects
  • Material offers good resistance to cuts and protection against thermal hazards (contact heat).
  • The length of the knitted wrist offers forearm protection
  • Provide protection for Level 2 heat exposure, which is effective up to 250°C
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-2003 Basic standards, EN 388-2016 Mechanical hazards, EN 407-2004 Thermical hazards (level 2)
Description 10 gauge
Color Yellow
Hand Ambidextrous
Material Dupont™ Kevlar™ Aramid Fibers
Size 7
Temperature (Metric) Max Effective up to 250°C
Liner Material Cotton
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell™ ARATHERMA FIT Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.