Learn More
STEMCELL Technologies ArciTect™ Human HPRT Positive Control Kit
Positive control for CRISPR-Cas9 genome editing
Brand:  STEMCELL Technologies 76013
Encompass_Preferred
Product Code. 17179301
  • £419.85 / 1 set
Explore more special offers
Description
Description
  • ArciTect™ Human HPRT Positive Control Kit is designed to enable optimization of  transfection protocols and act as a positive control for experiments using the ArciTect™ CRISPR-Cas9 genome editing system
  • The kit comprises ArciTect™ Human HPRT crRNA (2 nmol), ArciTect™ Human HPRT Primer Mix (2 nmol), ArciTect™ tracrRNA (5 nmol), and ArciTect™ Annealing Buffer (5X)
  • ArciTect™ Human HPRT crRNA first requires annealing to ArciTect™ tracrRNA
  • This must then be combined with an ArciTect™ Cas9 Nuclease (e.g. Catalog #76002) to form a ribonucleoprotein complex
Specifications
Specifications
Pluripotent Stem Cells
1 Kit