missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Argon Gloves
Offering flexibility, dexterity and mechanical resistance, these goat leather gloves are well-suited to the demands of welding. Honeywell Safety Products™ Argon Gloves are also designed with a split leather cuff for forearm protection.
£14.10 - £16.20
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Attestation Number 0075/797/162/11/06/0313 |
---|---|
Classification | Dexterity: 5 |
Cuff Material | Split leather |
Cut Resistance Rating | 1 |
Material | Grain Leather, Leather, Split Goat Leather |
Description
- Suitable for argon welding and fine handling.
- Goat leather grain glove construction offers ideal mechanical resistance for welders.
- Sewn-on, one-piece thumb.
- Good flexibility and dexterity.
- Split leather cuff protects the forearm.
Specifications
CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Attestation Number 0075/797/162/11/06/0313 | |
Split leather | |
Grain Leather, Leather, Split Goat Leather | |
0.9 to 1 mm | |
1 | |
Heat | |
35 cm |
Dexterity: 5 | |
1 | |
Welding Gloves | |
Automotive, Building, Construction and Welding industries | |
10Pair/PK | |
1 | |
15 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title