missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aripiprazole
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 149.0°C |
| Loss on Drying | 0.3% max. (105°C, 3 hrs) |
| Infrared Spectrum | Authentic |
| Quantity | 1 g |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in acetic acid, insoluble in methanol and hexane |
| Formula Weight | 448.39 |
| Percent Purity | ≥97.5% (HPLC) |
| Chemical Name or Material | Aripiprazole |
Safety and Handling
Product Identifier
- Aripiprazole
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Aripiprazole
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction