missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius arium™ comfort I ASTM 1, 3 Combined Water purification Systems with UV Lamp
Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.
£7960.00 - £7985.00
Specifications
Bacterial Content | <1 CFU/L |
---|---|
Daily Output | 120 L/hr. (Type 1), 8 L/hr. or 16 L/hr. (Type 3) |
Product Water Type | Type 1 and 3 |
Type | Water Purification System |
Feedwater Pressure | 2.0 to 6.9 bar |
Product Code | Brand | Flow Rate | Mounting | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Mounting | Price | Quantity & Availability | ||||
15584146
|
Sartorius
H2O-I-1-UV-T |
8 L/hr. (Type 3) | Benchtop |
£7985.00
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
15819354
|
Sartorius
H2O-I-1-UV-B |
8 L/hr. (Type 3) | Wall |
£7960.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
||||
Description
- Unique cartridge kit for production of highest quality ultrapure water
- Innovative iJust technology optimizes product water quality
- Touch display with intuitive menu navigation
- Compact design with SD card slot
- Reproducible and reliable results
- Arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
- Available as bench-top system for every laboratory workstation
- Wall-mounted with integrated wall bracket
Benefits
- Optimized, economic water consumption
- Premium product-water quality at all times
- Guaranteed longer life of downstream ultrapure water systems
Specifications
<1 CFU/L | |
Type 1 and 3 | |
2.0 to 6.9 bar | |
HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC-Analysis, Photometry, Buffer solutions, Feed water for laboratory devices, such as autoclaves, glassware washers etc | |
<1mL | |
<1 NTU | |
43.5 cm | |
50.1 cm | |
47.6 cm | |
Approx. 31 kg (Operating); 23 kg (Empty) |
120 L/hr. (Type 1), 8 L/hr. or 16 L/hr. (Type 3) | |
Water Purification System | |
2°C to 30°C | |
80% | |
Adsorption by spherical activated carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration | |
17 in. | |
19.7 in. | |
18.7 in. | |
68.3 lb. (Operating); 50.7 lb. (Empty) | |
100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.) |