missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ comfort II ASTM 1, 2 Combined Water purification Systems with UV Lamp
GREENER_CHOICEBIOPHARMA

Product Code. p-5039984 Shop All Sartorius Products
Click to view available options
Flow Rate:
10 L/hr.
5 L/hr.
Mounting:
Benchtop
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15594146

Brand: Sartorius H2OII1UVT

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.

  • Equipped with latest EDI technology and unique cartridge kit
  • Innovative iJust technology optimizes product water quality
  • Touch display with intuitive menu navigation
  • Compact design with SD card slot
  • Performs electrochemical deionization in third purification stage
  • Reliable and safe removal of contamination contained in feed water
  • arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
  • Available as bench-top system for every laboratory workstation and wall-mounted with integrated wall bracket

Benefits

  • Optimized, economic water consumption
  • Premium product-water quality at all times
  • Guaranteed longer life of downstream ultrapure water systems

To install arium™ comfort II as built-in unit for supplying Type II pure water at flow rates of 5L/hour, please order corresponding bench-top system and conversion kit H2O-ACK-D

TRUSTED_SUSTAINABILITY

Specifications

Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Daily Output 120 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 5 L/hr.
Product Water Type Type 1 and 2
Remote Dispenser No
Resistivity 18.2 Mohm.cm
Total Organic Carbon ≤2 ppb
Type Water Purification System
Feedwater Pressure 2.0 to 6.9 bar
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC Analysis, Photometry, Microbiological media and reagents, Histology, ELISA, RIA
Humidity Range 80%
Includes UV lamp
Mounting Benchtop
Particle Size <1mL
Purification Method Adsorption using spherical activated carbon, catalyst, reverse osmosis, electrochemical deionization, optional final particle and sterile filtration
Turbidity <1 NTU
Width (English) 17 in.
Width (Metric) 43.5 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 18.7 in.
Depth (Metric) 47.6 cm
Weight (English) 61.7 lb. (Empty); 79.3 lb. (Operating)
Weight (Metric) 28 kg (Empty); 36 kg (Operating)
Electrical Requirements 100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.