missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Arium™ Mini Extend Ultrapure Water System
Compact Arium™ Mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 L/day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.
£4270.00 - £6140.00
Specifications
Bacterial Content | <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge ) |
---|---|
Daily Output | 1 to 10 L |
Endotoxin Concentration | <0.001 EU/mL with a terminal ultrafiltration cartridge |
Feedwater Connector | 3/8 in. |
Feed Water Monitoring | No |
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
30057367
|
Sartorius
H2O-MAR-T |
Arium Mini Plus without UV |
£5500.00
1 pack |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
30057366
|
Sartorius
H2O-MAR-UV-T |
Arium Mini Plus with UV |
£6140.00
1 pack |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
30057368
|
Sartorius
H2O-MUR-T |
Arium Mini Essential without UV |
£4270.00
1 pack |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
30057365
|
Sartorius
H2O-MUR-UV-T |
Arium Mini Essential with UV |
£4920.00
1 pack |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
Description
- Flexible: Different versions for different inlet water and connection options, as well as the desired water withdrawal
- Compact: Space-saving with a width of only 28 cm (+ 9 cm for versions with attached dispensing unit)
- Intuitive: Color touch display, with direct access to all important information and dispensing options
- Innovative: Depending on the type of system, our unique Bagtank technology saves time as no intensive tank cleaning with hazardous substances is required
- Reliable: Delivers consistently high water quality for reliable and reproducible results
Specifications
<0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge ) | |
<0.001 EU/mL with a terminal ultrafiltration cartridge | |
No | |
Optional | |
Water Purification System |
1 to 10 L | |
3/8 in. | |
1 L/min. | |
18.2 MΩ x cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius Arium™ Mini Extend Ultrapure Water System