Learn More
Thermo Scientific™ Armadillo 96-well Clear PCR Plates, White Wells - PROMO
The ultimate in high performance PCR plastics combining the rigidity of a polycarbonate frame with thin walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping.
Brand: Thermo Scientific™ AB3596
81.88 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- Optically clear skirt—Aids in visualization of liquids and improves stacking capabilities in automated handling systems
- Raised rim design— Improved well surface area improves seal integrity in automated heat sealing
- ANSI:SBS footprint—With rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high throughput applications
- Thermostable polycarbonate semi-skirted frame design—Warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
- Thin walled U-bottom wells—Optimal heat transfer and maximum sample recovery
- Indented Alphanumeric Grid reference lettering—Integrated frame and well design for improved heat sealing
- Available in both clear and white colored wells—Ensures the highest level of sensitivity for all PCR/qPCR reactions
Specifications
No Barcode | |
8 x 12 | |
Real-Time PCR Systems or Thermal Cyclers | |
qPCR/Real-Time PCR | |
25 Plates | |
White |
Standard | |
Clear | |
96 | |
ANSI footprint ensures complete compatibility with robotic systems for high throughput applications | |
Raised |