missing translation for 'onlineSavingsMsg'
Learn More

ARNTL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ARNTL
Concentration 0.3 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16852834
View Documents
Proteintech
14268-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16842834
View Documents
Proteintech
14268-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

ARNTL
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Rat, Mouse
O00327, Q9EPW1, Q9WTL8
11865, 29657, 406
ARNTL Fusion Protein Ag5586
Primary
-20°C
ARNTL
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ARNT3, ARNTL, bHLH PAS protein JAP3, bHLHe5, BMAL1, BMAL1c, Brain and muscle ARNT like 1, JAP3, Member of PAS protein 3, MOP3, PASD3, TIC
ARNTL
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications