missing translation for 'onlineSavingsMsg'
Learn More

Arprinocid, Honeywell Fluka™

Product Code. 15940232
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 55779-18-5
Packaging Glass Bottle
Quantity 10 mg
Grade Analytical Standard
Density 1.59 g/cm3
Boiling Point 504°C
Flash Point 259°C
For Use With (Application) Coccidiostat
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'