Learn More
Thermo Scientific™ ArrayScan™ Liquid Handling Module
The Thermo Scientific™ ArrayScan™ Liquid Handling Module is designed to work with the Thermo Scientific™ ArrayScan™ Live Cell Module, expanding the flexibility of the platform for analysis of complex cellular states of cells.
Brand:  Thermo Scientific™ N010117B
Product Code. 15963763
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The ArrayScan Liquid Handling Module has functionalities to aspirate, dispense, and mix, from four deck locations. The module is capable of single-tip aspirate and dispense at any location, creating a highly flexible instrument. Compatible with 6-well, 8-well, 24-well, 96-well, and 384-well plates, the ArrayScan Liquid Handling Module utilizes disposable tips with a dispense volume range of 3 to 200μL.
Features of the ArrayScan Liquid Handling Module:
• Compatible with 6-well, 8-well, 24-well, 96-well, and 384-well plates, the module utilizes disposable tips with a dispense volume range of 3 to 200μL
• Full function liquid handling from any plate or trough (3 plate and 1 quarter trough)
• Industry standard arm with single pipette tip driven by syringe pump (e.g., aspirate, dispense)
• Compatible with all ArrayScan Modules
• Dynamic access to compound and assay plate
• One-to-many and many-to-one transfer scenarios are supported
Includes:
ArrayScan Liquid Handling Module : Automated, single-tip, liquid handling robot module for the Thermo Scientific™ ArrayScan™ High Content Analysis (HCA) Readers using disposable tips (standard and liquid level sensing) to both aspirate and dispense from multiple deck and live cell chamber locations.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ArrayScan
3 to 200 μL (± 5% in 1 μL increments)
Thermo Scientific™ HCS Studio Cell Analysis Software
87.63 cm (34.5 in.) (W) x 109.22 cm (43 in.) (H) x 74 cm (29 in.) (D)
1 each
150 W (+ 500 W from ArrayScan Reader)
ArrayScan HCA Readers
6-wells, 8-wells, 384-well plate, 24-well plate, 96-well plate
Permissible ambient temperature range during operation : + 18 deg C to + 25 deg C, Permissible relative humidity during operation (non-condensing): > 75% (at 30 deg C)
59 kg
100 V to 240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ ArrayScan™ Liquid Handling Module > 87.63 cm (34.5 in) (W) x 109.22 cm (43 in) (H) x 74 cm (29 in) (D)